Product Details

SNP ID
rs79691215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104986800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAAAAGAGTGCAGTTCTCACCA[A/G]CCATGCCAGGGTGGACCTTGGGTGA
Phenotype
MIM: 608570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHNAK2 PubMed Links

Gene Details

Gene
AHNAK2
Gene Name
AHNAK nucleoprotein 2
There are no transcripts associated with this gene.

Gene
C14orf79
Gene Name
chromosome 14 open reading frame 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174891.3 522 Silent Mutation CAA,CAG Q123Q NP_777551.2
XM_005267317.3 522 Missense Mutation CAA,CAG Q123Q XP_005267374.1
XM_005267318.3 522 Missense Mutation CAA,CAG Q123Q XP_005267375.1

View Full Product Details