Product Details

SNP ID
rs78360052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48568587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCATCAGTAATGTCATCATCCA[C/G]GAACTCTGGCAGGAGTTACAGGGAA
Phenotype
MIM: 149750
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LALBA PubMed Links

Gene Details

Gene
LALBA
Gene Name
lactalbumin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002289.2 402 Missense Mutation CTG,GTG L100V NP_002280.1
XM_006719395.3 402 Missense Mutation CTG,GTG L107V XP_006719458.1

View Full Product Details