Product Details

SNP ID
rs79571226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:80132050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAATGTACATTCTCTGCCCCCT[C/T]AACAGACCTACTGAATCAAAAACTC
Phenotype
MIM: 614925
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTOGL PubMed Links

Gene Details

Gene
OTOGL
Gene Name
otogelin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173591.3 582 Intron NP_775862.3
XM_005268802.3 582 UTR 5 XP_005268859.1
XM_011538191.1 582 Intron XP_011536493.1
XM_011538192.2 582 Intron XP_011536494.1
XM_011538193.1 582 Intron XP_011536495.1

View Full Product Details