Product Details

SNP ID
rs79397427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42160808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAATGCTGAGCACTACTCCGATC[A/C]ACATTTTTCAATCTTGGCCTAAACA
Phenotype
MIM: 607534
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
YAF2 PubMed Links

Gene Details

Gene
YAF2
Gene Name
YY1 associated factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190977.2 512 Silent Mutation GTG,GTT V66V NP_001177906.1
NM_001190979.2 512 Silent Mutation GTG,GTT V132V NP_001177908.1
NM_001190980.2 512 Intron NP_001177909.1
NM_001320080.1 512 Silent Mutation GTG,GTT V99V NP_001307009.1
NM_005748.5 512 Silent Mutation GTG,GTT V108V NP_005739.2
XM_006719184.3 512 Silent Mutation GTG,GTT V110V XP_006719247.1
XM_006719185.3 512 Silent Mutation GTG,GTT V105V XP_006719248.1
XM_011537728.2 512 Silent Mutation GTG,GTT V166V XP_011536030.1
XM_011537729.2 512 Silent Mutation GTG,GTT V87V XP_011536031.1
XM_017018669.1 512 Silent Mutation GTG,GTT V115V XP_016874158.1
XM_017018670.1 512 Intron XP_016874159.1

View Full Product Details