Product Details

SNP ID
rs77270708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42164120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAATGGTTGCCTAGTTAATAAA[C/T]GACTACTGTCTAGCCACTGGCCAAC
Phenotype
MIM: 607534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YAF2 PubMed Links

Gene Details

Gene
YAF2
Gene Name
YY1 associated factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190977.2 Intron NP_001177906.1
NM_001190979.2 Intron NP_001177908.1
NM_001190980.2 Intron NP_001177909.1
NM_001320080.1 Intron NP_001307009.1
NM_005748.5 Intron NP_005739.2
XM_006719184.3 Intron XP_006719247.1
XM_006719185.3 Intron XP_006719248.1
XM_011537728.2 Intron XP_011536030.1
XM_011537729.2 Intron XP_011536031.1
XM_017018669.1 Intron XP_016874158.1
XM_017018670.1 Intron XP_016874159.1

View Full Product Details