Product Details

SNP ID
rs79907632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85286918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGTTATGGCCAAATACAACAAGC[A/G]AAAAGCCATTTTGCTGCCACCTATG
Phenotype
MIM: 601527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALX1 PubMed Links

Gene Details

Gene
ALX1
Gene Name
ALX homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006982.2 626 Silent Mutation GCA,GCG A199A NP_008913.2
XM_011538782.2 626 Silent Mutation GCA,GCG A104A XP_011537084.1

View Full Product Details