Product Details

SNP ID
rs78431175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85973411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGGTTAAAGCTGTAGTAACTTA[C/T]AGTACATTGAAATTATAGACTAAGG
Phenotype
MIM: 607385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGAT4C PubMed Links

Gene Details

Gene
MGAT4C
Gene Name
MGAT4 family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013244.3 8307 Intron NP_037376.2
XM_005268776.4 8307 Intron XP_005268833.1
XM_005268779.3 8307 UTR 3 XP_005268836.1
XM_011538150.2 8307 Intron XP_011536452.1
XM_011538151.2 8307 Intron XP_011536453.1
XM_011538156.2 8307 UTR 3 XP_011536458.1
XM_011538157.2 8307 UTR 3 XP_011536459.1
XM_017019143.1 8307 Intron XP_016874632.1
XM_017019144.1 8307 Intron XP_016874633.1
XM_017019145.1 8307 Intron XP_016874634.1
XM_017019146.1 8307 Intron XP_016874635.1
XM_017019147.1 8307 Intron XP_016874636.1
XM_017019148.1 8307 UTR 3 XP_016874637.1
XM_017019149.1 8307 UTR 3 XP_016874638.1
XM_017019150.1 8307 UTR 3 XP_016874639.1

View Full Product Details