Product Details

SNP ID
rs76053767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95078029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCCTGTTTTCAATTGAACAAAT[A/G]TTTTCTCTTTAAAGACCCTCCTAGG
Phenotype
MIM: 613520 MIM: 601529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGD6 PubMed Links

Gene Details

Gene
FGD6
Gene Name
FYVE, RhoGEF and PH domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018351.3 8008 UTR 3 NP_060821.3
Gene
NR2C1
Gene Name
nuclear receptor subfamily 2 group C member 1
There are no transcripts associated with this gene.

View Full Product Details