Product Details

SNP ID
rs76298705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:43361058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTACTTTCGACATATGCATTC[C/T]AAAGTCACACCACACTCCTTAACTT
Phenotype
MIM: 611681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS20 PubMed Links

Gene Details

Gene
ADAMTS20
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025003.3 Intron NP_079279.3
XM_011538754.2 Intron XP_011537056.1
XM_017019979.1 Intron XP_016875468.1

View Full Product Details