Product Details

SNP ID
rs74366368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103990375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCTACTTTTTAAACAAACAGCT[A/G]TCTATCTCCACCTGAACATTACTCC
Phenotype
MIM: 601423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLT8D2 PubMed Links

Gene Details

Gene
GLT8D2
Gene Name
glycosyltransferase 8 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316967.1 Intron NP_001303896.1
NM_031302.4 Intron NP_112592.1
XM_011538793.2 Intron XP_011537095.1
XM_017019999.1 Intron XP_016875488.1
Gene
TDG
Gene Name
thymine DNA glycosylase
There are no transcripts associated with this gene.

View Full Product Details