Product Details

SNP ID
rs79545524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106956985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAACACCTAGAGAAAAGGGAAG[A/G]GTTCTTAATTTGATTTTTGCGTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM263 PubMed Links

Gene Details

Gene
TMEM263
Gene Name
transmembrane protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319661.1 201 Intron NP_001306590.1
NM_001319662.1 201 UTR 5 NP_001306591.1
NM_001319663.1 201 Intron NP_001306592.1
NM_001319664.1 201 Intron NP_001306593.1
NM_001319666.1 201 Intron NP_001306595.1
NM_152261.3 201 Intron NP_689474.1
XM_017020198.1 201 Intron XP_016875687.1

View Full Product Details