Product Details
- SNP ID
-
rs77583795
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:107686502 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAGTAATCCAAGTGAGATGTTG[A/T]TGGTAGCAGGGAGGATGAGAGGTAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PWP1
PubMed Links
Gene Details
- Gene
- PWP1
- Gene Name
- PWP1 homolog, endonuclein
View Full Product Details