Product Details

SNP ID
rs75283310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108648836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTGTGTCAGGATACAGGTCATC[C/T]TGGAAAAGGTCAGACTACAAGAGAC
Phenotype
MIM: 605269
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO1C PubMed Links

Gene Details

Gene
CORO1C
Gene Name
coronin 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105237.2 1282 Silent Mutation CAA,CAG Q411Q NP_001098707.1
NM_001276471.1 1282 Silent Mutation CAA,CAG Q358Q NP_001263400.1
NM_014325.3 1282 Silent Mutation CAA,CAG Q358Q NP_055140.1
XM_011538125.1 1282 Silent Mutation CAA,CAG Q170Q XP_011536427.1
XM_017019121.1 1282 Silent Mutation CAA,CAG Q411Q XP_016874610.1
XM_017019122.1 1282 Silent Mutation CAA,CAG Q358Q XP_016874611.1
XM_017019123.1 1282 Silent Mutation CAA,CAG Q358Q XP_016874612.1

View Full Product Details