Product Details

SNP ID
rs78832270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196646066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCGCCCACTAAACGTGAAGTCC[A/G]AACATTCCCTTACATCGACAAAGCC
Phenotype
MIM: 615322
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRROS PubMed Links

Gene Details

Gene
NRROS
Gene Name
negative regulator of reactive oxygen species
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198565.2 Intron NP_940967.1

View Full Product Details