Product Details

SNP ID
rs77792747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197963905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTAAGTATGATGTTAACTTTAC[A/G]TTTTTTTGTAGTTCCCTTTATCAGA
Phenotype
MIM: 612477 MIM: 609380 MIM: 180468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQCG PubMed Links

Gene Details

Gene
IQCG
Gene Name
IQ motif containing G
There are no transcripts associated with this gene.

Gene
LMLN
Gene Name
leishmanolysin like peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136049.2 Intron NP_001129521.2
NM_033029.3 Intron NP_149018.2
XM_011513270.1 Intron XP_011511572.1
XM_011513271.1 Intron XP_011511573.1
XM_017007428.1 Intron XP_016862917.1
Gene
RPL35A
Gene Name
ribosomal protein L35a
There are no transcripts associated with this gene.

View Full Product Details