Product Details

SNP ID
rs79554134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68956270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACGCTGGTTTGGAATCAGACAC[C/T]GCAATGAAAAAAGGGGAGACACTGC
Phenotype
MIM: 606357 MIM: 610373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX21 PubMed Links

Gene Details

Gene
DDX21
Gene Name
DEAD-box helicase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256910.1 4364 Intron NP_001243839.1
NM_004728.3 4364 Silent Mutation ACC,ACT T15T NP_004719.2
XM_011540336.1 4364 Silent Mutation ACC,ACT T15T XP_011538638.1
XM_017016910.1 4364 Intron XP_016872399.1
Gene
DDX50
Gene Name
DEAD-box helicase 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024045.1 4364 Intron NP_076950.1
XM_005270148.2 4364 Intron XP_005270205.1
XM_011540144.2 4364 Intron XP_011538446.1
XM_017016626.1 4364 UTR 3 XP_016872115.1
XM_017016627.1 4364 Intron XP_016872116.1
XM_017016628.1 4364 Intron XP_016872117.1
XM_017016629.1 4364 Intron XP_016872118.1
XM_017016630.1 4364 Intron XP_016872119.1

View Full Product Details