Product Details

SNP ID
rs74903725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84194804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCCCTGGCCCTGGGGCTGCTGC[G/T]CCTCTGCTTGGGTGAGTGGCCGCTG
Phenotype
MIM: 609502
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf99 PubMed Links

Gene Details

Gene
C10orf99
Gene Name
chromosome 10 open reading frame 99
There are no transcripts associated with this gene.

Gene
CDHR1
Gene Name
cadherin related family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171971.2 170 Missense Mutation CGC,CTC R15L NP_001165442.1
NM_033100.3 170 Missense Mutation CGC,CTC R15L NP_149091.1
XM_011540337.1 170 Intron XP_011538639.1
XM_011540338.1 170 Intron XP_011538640.1
XM_011540339.1 170 UTR 5 XP_011538641.1
XM_011540340.2 170 Intron XP_011538642.1

View Full Product Details