Product Details

SNP ID
rs78209918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88978672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCCAGGCTCAGTTCATTTGCAC[C/T]ACGGCTCCAGCTCACCCTACTTCAG
Phenotype
MIM: 102620 MIM: 134637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTA2 PubMed Links

Gene Details

Gene
ACTA2
Gene Name
actin, alpha 2, smooth muscle, aorta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141945.2 Intron NP_001135417.1
NM_001320855.1 Intron NP_001307784.1
NM_001613.2 Intron NP_001604.1
Gene
FAS
Gene Name
Fas cell surface death receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000043.5 Intron NP_000034.1
NM_001320619.1 Intron NP_001307548.1
NM_152871.3 Intron NP_690610.1
NM_152872.3 Intron NP_690611.1
XM_006717819.3 Intron XP_006717882.1
XM_011539764.2 Intron XP_011538066.1
XM_011539765.2 Intron XP_011538067.1
XM_011539766.2 Intron XP_011538068.1
XM_011539767.2 Intron XP_011538069.1

View Full Product Details