Product Details
- SNP ID
-
rs75426652
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124461954 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACGACAGCGGCGCGGGCGGCGGCA[A/C]GGCCATCGCCGGCTCGGTGGAGGCG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LHPP
PubMed Links
Gene Details
- Gene
- LHPP
- Gene Name
- phospholysine phosphohistidine inorganic pyrophosphate phosphatase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001167880.1 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
NP_001161352.1 |
NM_001318331.1 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
NP_001305260.1 |
NM_001318332.1 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
NP_001305261.1 |
NM_022126.3 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
NP_071409.3 |
XM_005270026.3 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
XP_005270083.1 |
XM_006717943.1 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
XP_006718006.1 |
XM_011540058.2 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
XP_011538360.1 |
XM_017016509.1 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
XP_016871998.1 |
XM_017016510.1 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
XP_016871999.1 |
XM_017016511.1 |
183 |
Missense Mutation |
AAG,ACG |
K31T |
XP_016872000.1 |
XM_017016512.1 |
183 |
Intron |
|
|
XP_016872001.1 |
View Full Product Details