Product Details

SNP ID
rs76006730
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:52427419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGTTTGTGGGACATTAATTAGT[C/T]CACTATGACTAAAACACTGAGAACA
Phenotype
MIM: 617031 MIM: 613692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPF38A PubMed Links

Gene Details

Gene
PRPF38A
Gene Name
pre-mRNA processing factor 38A
There are no transcripts associated with this gene.

Gene
ZCCHC11
Gene Name
zinc finger CCHC-type containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009881.2 Intron NP_001009881.1
NM_015269.2 Intron NP_056084.1
XM_005270676.2 Intron XP_005270733.1
XM_005270678.2 Intron XP_005270735.1
XM_005270679.2 Intron XP_005270736.1
XM_005270680.2 Intron XP_005270737.1
XM_005270681.2 Intron XP_005270738.1
XM_005270682.2 Intron XP_005270739.1
XM_005270683.2 Intron XP_005270740.1
XM_006710498.2 Intron XP_006710561.1
XM_006710499.3 Intron XP_006710562.1
XM_006710500.1 Intron XP_006710563.1
XM_011541097.1 Intron XP_011539399.1
XM_011541098.1 Intron XP_011539400.1
XM_011541099.1 Intron XP_011539401.1
XM_011541100.2 Intron XP_011539402.1
XM_011541101.2 Intron XP_011539403.1
XM_017000803.1 Intron XP_016856292.1
XM_017000804.1 Intron XP_016856293.1
XM_017000805.1 Intron XP_016856294.1
XM_017000806.1 Intron XP_016856295.1
XM_017000807.1 Intron XP_016856296.1
XM_017000808.1 Intron XP_016856297.1
XM_017000809.1 Intron XP_016856298.1
XM_017000810.1 Intron XP_016856299.1
XM_017000811.1 Intron XP_016856300.1
XM_017000812.1 Intron XP_016856301.1
XM_017000813.1 Intron XP_016856302.1
XM_017000814.1 Intron XP_016856303.1
XM_017000815.1 Intron XP_016856304.1
XM_017000816.1 Intron XP_016856305.1
XM_017000817.1 Intron XP_016856306.1
XM_017000818.1 Intron XP_016856307.1

View Full Product Details