Product Details

SNP ID
rs79299252
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:54851289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCCACTTTCCCACCTGATGCT[A/G]TCCACTCCCCAGCCCCTGCTGCAGA
Phenotype
MIM: 606418 MIM: 616446
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHCR24 PubMed Links

Gene Details

Gene
DHCR24
Gene Name
24-dehydrocholesterol reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014762.3 2624 UTR 3 NP_055577.1
Gene
LEXM
Gene Name
lymphocyte expansion molecule
There are no transcripts associated with this gene.

View Full Product Details