Product Details

SNP ID
rs75390249
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:63774030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAAGTGACAAGTTGAGCGAGA[A/G]AGGGAGCGTGGAGAGCTGGAGCAGC
Phenotype
MIM: 602336
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ROR1 PubMed Links

Gene Details

Gene
ROR1
Gene Name
receptor tyrosine kinase like orphan receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083592.1 12 UTR 5 NP_001077061.1
NM_005012.3 12 UTR 5 NP_005003.2
XM_011541526.1 12 Intron XP_011539828.1
XM_017001376.1 12 Intron XP_016856865.1
XM_017001377.1 12 Intron XP_016856866.1

View Full Product Details