Product Details

SNP ID
rs77435924
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:70262269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCTCGAAGGAATATTTATAAA[C/T]GTCACCTTGCATAAAGATGACAAGT
Phenotype
MIM: 615125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13C PubMed Links

Gene Details

Gene
ANKRD13C
Gene Name
ankyrin repeat domain 13C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030816.4 1369 UTR 3 NP_110443.3
XM_005271234.3 1369 UTR 3 XP_005271291.1
XM_005271235.3 1369 UTR 3 XP_005271292.1
XM_006710929.3 1369 UTR 3 XP_006710992.1
XM_017002413.1 1369 UTR 3 XP_016857902.1
XM_017002414.1 1369 UTR 3 XP_016857903.1
XM_017002415.1 1369 UTR 3 XP_016857904.1
XM_017002416.1 1369 UTR 3 XP_016857905.1

View Full Product Details