Product Details

SNP ID
rs74637079
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:74027726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAGGAGATTTCATTCATTCATC[C/T]TTATTTATTCAACAATTATGTATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRIQ3 PubMed Links

Gene Details

Gene
LRRIQ3
Gene Name
leucine rich repeats and IQ motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105659.1 Intron NP_001099129.1
NM_001322315.1 Intron NP_001309244.1
XM_011540658.1 Intron XP_011538960.1
XM_011540659.1 Intron XP_011538961.1
XM_011540660.1 Intron XP_011538962.1
XM_011540662.2 Intron XP_011538964.1
XM_011540663.2 Intron XP_011538965.1
XM_017000286.1 Intron XP_016855775.1
XM_017000287.1 Intron XP_016855776.1
XM_017000288.1 Intron XP_016855777.1

View Full Product Details