Product Details

SNP ID
rs76669666
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:76082558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGGATTATCTGCGCTTTCTCT[A/G]TTGCTGTTTCTCTCAATTCAAAAAT
Phenotype
MIM: 610133
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST6GALNAC3 PubMed Links

Gene Details

Gene
ST6GALNAC3
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160011.1 Intron NP_001153483.1
NM_152996.2 Intron NP_694541.2
XM_006710553.3 Intron XP_006710616.1
XM_017000937.1 Intron XP_016856426.1
XM_017000938.1 Intron XP_016856427.1
XM_017000939.1 Intron XP_016856428.1
XM_017000940.1 Intron XP_016856429.1
XM_017000941.1 Intron XP_016856430.1
XM_017000942.1 Intron XP_016856431.1

View Full Product Details