Product Details

SNP ID
rs76038246
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:78046785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATCTCCCTAGATTAACGGGGG[A/G]AAAAAAAAATAGAAGTGCCTTGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIPC2 PubMed Links

Gene Details

Gene
GIPC2
Gene Name
GIPC PDZ domain containing family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304725.1 Intron NP_001291654.1
NM_017655.5 Intron NP_060125.4

View Full Product Details