Product Details

SNP ID
rs78911400
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:91957670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTAATAGTTTCCATAGTTTCTG[C/T]TTTGGTGTACATTCCATGAGTTTTG
Phenotype
MIM: 602144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRDT PubMed Links

Gene Details

Gene
BRDT
Gene Name
bromodomain testis associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242805.2 Intron NP_001229734.2
NM_001242806.2 Intron NP_001229735.2
NM_001242807.2 Intron NP_001229736.2
NM_001242808.2 Intron NP_001229737.2
NM_001242810.2 Intron NP_001229739.2
NM_001726.4 Intron NP_001717.3
NM_207189.3 Intron NP_997072.2
XM_006710853.3 Intron XP_006710916.1
XM_006710854.3 Intron XP_006710917.1
XM_006710855.3 Intron XP_006710918.1
XM_006710856.3 Intron XP_006710919.1
XM_006710857.3 Intron XP_006710920.1
XM_011542032.2 Intron XP_011540334.1
XM_011542033.2 Intron XP_011540335.1
XM_011542034.2 Intron XP_011540336.1
XM_011542035.2 Intron XP_011540337.1
XM_011542036.2 Intron XP_011540338.1

View Full Product Details