Product Details

SNP ID
rs77293072
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:93996193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTGTTTAGCAAAATTTACAAA[C/T]ACCTAGAGGTAAGAGAAGAGCGAGA
Phenotype
MIM: 601691
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCA4 PubMed Links

Gene Details

Gene
ABCA4
Gene Name
ATP binding cassette subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000350.2 6836 Silent Mutation GTA,GTG V2244V NP_000341.2

View Full Product Details