Product Details

SNP ID
rs80168198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:115177362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACTCTTATCACCAAATCCCAA[C/T]AGTTTACATGGAGAACAACAAAGAC
Phenotype
MIM: 605686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CADM1 PubMed Links

Gene Details

Gene
CADM1
Gene Name
cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098517.1 Intron NP_001091987.1
NM_001301043.1 Intron NP_001287972.1
NM_001301044.1 Intron NP_001287973.1
NM_001301045.1 Intron NP_001287974.1
NM_014333.3 Intron NP_055148.3
XM_005271494.2 Intron XP_005271551.1
XM_017017457.1 Intron XP_016872946.1
XM_017017458.1 Intron XP_016872947.1
XM_017017459.1 Intron XP_016872948.1
XM_017017460.1 Intron XP_016872949.1
XM_017017461.1 Intron XP_016872950.1

View Full Product Details