Product Details

SNP ID
rs76771914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123805762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGACAAGGGCAGAGAGGAGAAA[A/G]TTTATCTTCTCAATGAGGTGAGTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6M1 PubMed Links

Gene Details

Gene
OR6M1
Gene Name
olfactory receptor family 6 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005325.1 588 Silent Mutation AAC,AAT N196N NP_001005325.1

View Full Product Details