Product Details

SNP ID
rs77342042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111500000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGTCTGAATATATCCTGTCAA[A/G]AGACTCTGGTTTGGGTTGTCTTTAA
Phenotype
MIM: 114080 MIM: 607049
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMK4 PubMed Links

Gene Details

Gene
CAMK4
Gene Name
calcium/calmodulin dependent protein kinase IV
There are no transcripts associated with this gene.

Gene
STARD4
Gene Name
StAR related lipid transfer domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308056.1 726 Silent Mutation CTC,CTT L168L NP_001294985.1
NM_001308057.1 726 Intron NP_001294986.1
NM_001308058.1 726 Intron NP_001294987.1
NM_001308059.1 726 Silent Mutation CTC,CTT L86L NP_001294988.1
NM_001308060.1 726 Silent Mutation CTC,CTT L70L NP_001294989.1
NM_001308061.1 726 Silent Mutation CTC,CTT L86L NP_001294990.1
NM_139164.2 726 Silent Mutation CTC,CTT L168L NP_631903.1
XM_017009041.1 726 Silent Mutation CTC,CTT L70L XP_016864530.1
XM_017009042.1 726 Silent Mutation CTC,CTT L70L XP_016864531.1
XM_017009043.1 726 Silent Mutation CTC,CTT L70L XP_016864532.1
XM_017009044.1 726 Silent Mutation CTC,CTT L86L XP_016864533.1

View Full Product Details