Product Details
- SNP ID
-
rs77688129
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:112157110 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGGAGAGACAAATAGACCAGTAG[A/G]ACAGAATAGAGAGTGCAAAGGTACA
- Phenotype
-
MIM: 612141
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EPB41L4A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs376822884] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPB41L4A
- Gene Name
- erythrocyte membrane protein band 4.1 like 4A
- Gene
- EPB41L4A-AS1
- Gene Name
- EPB41L4A antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- SNORA13
- Gene Name
- small nucleolar RNA, H/ACA box 13
There are no transcripts associated with this gene.
View Full Product Details