Product Details

SNP ID
rs79192921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:113023952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAATCTTGATCGGGTAACTACGG[C/T]GCACTTTTTTCTCATTTAAAATAAT
Phenotype
MIM: 609844 MIM: 159350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCP2 PubMed Links

Gene Details

Gene
DCP2
Gene Name
decapping mRNA 2
There are no transcripts associated with this gene.

Gene
MCC
Gene Name
mutated in colorectal cancers
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085377.1 6470 UTR 3 NP_001078846.1
NM_002387.2 6470 UTR 3 NP_002378.1
XM_005271991.2 6470 UTR 3 XP_005272048.1
XM_017009473.1 6470 UTR 3 XP_016864962.1
XM_017009474.1 6470 UTR 3 XP_016864963.1

View Full Product Details