Product Details

SNP ID
rs74755145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:136213991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAACCCCATTAACCAGAGCATTG[A/C]TTTTGATGATGCTATGTTTTGTTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRPC7 PubMed Links

Gene Details

Gene
TRPC7
Gene Name
transient receptor potential cation channel subfamily C member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167576.1 Intron NP_001161048.1
NM_001167577.1 Intron NP_001161049.1
NM_020389.2 Intron NP_065122.1

View Full Product Details