Product Details

SNP ID
rs80086291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138253352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGAAGAGTTGGCTGTGAAAAC[G/A]CATCTTAGCTGCAATGGCCTCCGCT
Phenotype
MIM: 605710
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GFRA3 PubMed Links

Gene Details

Gene
GFRA3
Gene Name
GDNF family receptor alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001496.3 1188 Missense Mutation CGT,TGT R350C NP_001487.2

View Full Product Details