Product Details

SNP ID
rs75067698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:959671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACATCAACAGCGGGCAAGCAGC[A/G]TGGAAAGTGCTAGCGTGGGGTCTGG
Phenotype
MIM: 601207 MIM: 616660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKQ PubMed Links

Gene Details

Gene
DGKQ
Gene Name
diacylglycerol kinase theta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001347.3 3841 UTR 3 NP_001338.2
XM_011513411.1 3841 UTR 3 XP_011511713.1
XM_011513412.1 3841 Intron XP_011511714.1
XM_011513414.2 3841 Intron XP_011511716.1
XM_011513415.1 3841 Intron XP_011511717.1
XM_017007814.1 3841 Intron XP_016863303.1
XM_017007815.1 3841 Intron XP_016863304.1
Gene
TMEM175
Gene Name
transmembrane protein 175
There are no transcripts associated with this gene.

View Full Product Details