Product Details
- SNP ID
-
rs79363216
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:227817079 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACAGCCAGATAGGTCCCTACTA[C/T]GCTGCAGGAAGAAACTGGCTTTTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS38
PubMed Links
Gene Details
- Gene
- PRSS38
- Gene Name
- protease, serine 38
View Full Product Details