Product Details

SNP ID
rs74441001
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:244835450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCGGTTTGTTTTCCGAACCCAT[C/T]TCCTTCCGCTCTCCCGCCCCTGCAG
Phenotype
MIM: 614698 MIM: 602869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX20 PubMed Links

Gene Details

Gene
COX20
Gene Name
COX20, cytochrome c oxidase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312871.1 122 Intron NP_001299800.1
NM_001312872.1 122 Intron NP_001299801.1
NM_001312873.1 122 Intron NP_001299802.1
NM_001312874.1 122 Intron NP_001299803.1
NM_198076.5 122 UTR 5 NP_932342.1
Gene
HNRNPU
Gene Name
heterogeneous nuclear ribonucleoprotein U
There are no transcripts associated with this gene.

View Full Product Details