Product Details
- SNP ID
-
rs77223856
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
19
- Location
-
Chr.1:248295483 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTGCTAAACAGGGAGGTCAAAAC[G/T]GTGGCCAGAAGCATCATGAAGAGGA
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2T12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12135684] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2T12
- Gene Name
- olfactory receptor family 2 subfamily T member 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004692.1 |
96 |
Silent Mutation |
ACA,ACC |
T32T |
NP_001004692.1 |
View Full Product Details