Product Details

SNP ID
rs76663550
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:211262722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCATTGATCTGATCATTGGAAA[A/G]CATCTTTTTTGTTTCAGGATAAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RCOR3 PubMed Links

Gene Details

Gene
RCOR3
Gene Name
REST corepressor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136223.1 Intron NP_001129695.1
NM_001136224.2 Intron NP_001129696.2
NM_001136225.1 Intron NP_001129697.1
NM_018254.3 Intron NP_060724.1
XM_005273191.4 Intron XP_005273248.1
XM_006711442.2 Intron XP_006711505.1
XM_011509748.2 Intron XP_011508050.1
XM_011509749.2 Intron XP_011508051.1
XM_017001758.1 Intron XP_016857247.1
XM_017001759.1 Intron XP_016857248.1
XM_017001760.1 Intron XP_016857249.1
XM_017001761.1 Intron XP_016857250.1
XM_017001762.1 Intron XP_016857251.1
XM_017001763.1 Intron XP_016857252.1
XM_017001764.1 Intron XP_016857253.1
XM_017001765.1 Intron XP_016857254.1
XM_017001766.1 Intron XP_016857255.1
XM_017001767.1 Intron XP_016857256.1

View Full Product Details