Product Details

SNP ID
rs78551780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67129279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCATTTAGAACAAATAATAAAG[C/T]GTCTAGTTGATTTTAAAAGTTTAGT
Phenotype
MIM: 605657
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM2A PubMed Links

Gene Details

Gene
KDM2A
Gene Name
lysine demethylase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256405.1 Intron NP_001243334.1
NM_012308.2 Intron NP_036440.1
XM_006718480.3 Intron XP_006718543.1
XM_011544860.2 Intron XP_011543162.1
XM_011544861.1 Intron XP_011543163.1
XM_017017391.1 Intron XP_016872880.1
XM_017017392.1 Intron XP_016872881.1

View Full Product Details