Product Details

SNP ID
rs78822964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67292172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCTGGACACTCGTAATGTGGC[C/G]TTTGAGAGGTCGGTCGGGTCCTGGC
Phenotype
MIM: 615126 MIM: 109635
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD13D PubMed Links

Gene Details

Gene
ANKRD13D
Gene Name
ankyrin repeat domain 13D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207354.2 156 Silent Mutation GCC,GCG A241A NP_997237.2
XM_011544985.1 156 Silent Mutation GCC,GCG A241A XP_011543287.1
XM_011544986.1 156 Silent Mutation GCC,GCG A154A XP_011543288.1
XM_011544987.2 156 Silent Mutation GCC,GCG A154A XP_011543289.1
XM_011544988.2 156 UTR 5 XP_011543290.1
Gene
GRK2
Gene Name
G protein-coupled receptor kinase 2
There are no transcripts associated with this gene.

View Full Product Details