Product Details

SNP ID
rs75480027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74843815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCCTTTAGCCAGCAGGCGTGA[A/T]CTGGATCTGGGATCCTGGCAGCTGT
Phenotype
MIM: 609788
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF169 PubMed Links

Gene Details

Gene
RNF169
Gene Name
ring finger protein 169
There are no transcripts associated with this gene.

Gene
XRRA1
Gene Name
X-ray radiation resistance associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270380.1 2520 Intron NP_001257309.1
NM_001270381.1 2520 Intron NP_001257310.1
NM_182969.2 2520 Intron NP_892014.1
XM_005273765.2 2520 Intron XP_005273822.1
XM_005273770.2 2520 Intron XP_005273827.1
XM_011544755.1 2520 Intron XP_011543057.1
XM_011544756.1 2520 Intron XP_011543058.1
XM_011544757.1 2520 Intron XP_011543059.1
XM_011544758.1 2520 Intron XP_011543060.1
XM_011544759.1 2520 Intron XP_011543061.1
XM_011544760.2 2520 Missense Mutation ATC,TTC I730F XP_011543062.1
XM_011544761.1 2520 Intron XP_011543063.1
XM_011544762.1 2520 Intron XP_011543064.1
XM_011544763.1 2520 Intron XP_011543065.1
XM_011544764.1 2520 Intron XP_011543066.1
XM_011544765.1 2520 Intron XP_011543067.1
XM_011544766.2 2520 Intron XP_011543068.1
XM_011544767.1 2520 Intron XP_011543069.1
XM_011544768.1 2520 Intron XP_011543070.1
XM_017017221.1 2520 Intron XP_016872710.1
XM_017017222.1 2520 Intron XP_016872711.1
XM_017017223.1 2520 Intron XP_016872712.1
XM_017017224.1 2520 Intron XP_016872713.1
XM_017017225.1 2520 Intron XP_016872714.1
XM_017017226.1 2520 Intron XP_016872715.1
XM_017017227.1 2520 Intron XP_016872716.1
XM_017017228.1 2520 Intron XP_016872717.1
XM_017017229.1 2520 Intron XP_016872718.1
XM_017017230.1 2520 Intron XP_016872719.1
XM_017017231.1 2520 Intron XP_016872720.1
XM_017017232.1 2520 Intron XP_016872721.1
XM_017017233.1 2520 Intron XP_016872722.1
XM_017017234.1 2520 Intron XP_016872723.1

View Full Product Details