Product Details

SNP ID
rs78459567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57730834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGAGAGACATTTAGGGTAAAAC[A/G]GCCTAACCATTGTGAATGATCTATG
Phenotype
MIM: 616715
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMX2 PubMed Links

Gene Details

Gene
TMX2
Gene Name
thioredoxin related transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144012.2 Intron NP_001137484.1
NM_015959.3 Intron NP_057043.1
Gene
TMX2-CTNND1
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details