Product Details

SNP ID
rs78912817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57763937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGTTAACCTAGTGTCCAGGACT[A/G]TTTCCACGAACTAAGGGTTGTTTTG
Phenotype
MIM: 601045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTNND1 PubMed Links

Gene Details

Gene
CTNND1
Gene Name
catenin delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085458.1 Intron NP_001078927.1
NM_001085459.1 Intron NP_001078928.1
NM_001085460.1 Intron NP_001078929.1
NM_001085461.1 Intron NP_001078930.1
NM_001085462.1 Intron NP_001078931.1
NM_001085463.1 Intron NP_001078932.1
NM_001085464.1 Intron NP_001078933.1
NM_001085465.1 Intron NP_001078934.1
NM_001085466.1 Intron NP_001078935.1
NM_001085467.1 Intron NP_001078936.1
NM_001085468.1 Intron NP_001078937.1
NM_001085469.1 Intron NP_001078938.1
NM_001206883.1 Intron NP_001193812.1
NM_001206884.1 Intron NP_001193813.1
NM_001206885.1 Intron NP_001193814.1
NM_001206886.1 Intron NP_001193815.1
NM_001206887.1 Intron NP_001193816.1
NM_001206888.1 Intron NP_001193817.1
NM_001206889.1 Intron NP_001193818.1
NM_001206890.1 Intron NP_001193819.1
NM_001206891.1 Intron NP_001193820.1
NM_001331.2 Intron NP_001322.1
Gene
TMX2-CTNND1
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details