Product Details
- SNP ID
-
rs115484371
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:10065924 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTACTCAGAGACATTGCCGCTCCC[C/T]GGGTGGCCCTCTGTGCTCTGCTCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYS1
PubMed Links
Gene Details
- Gene
- CYS1
- Gene Name
- cystin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037160.2 |
539 |
Silent Mutation |
CCA,CCG |
P117P |
NP_001032237.1 |
View Full Product Details