Product Details
- SNP ID
-
rs114874345
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:12723490 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCCTCGGCAGTGGCCCACTGCCA[C/T]GACGGGGGGCTGGTGCTGCGGGACC
- Phenotype
-
MIM: 609462
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRIB2
PubMed Links
Gene Details
- Gene
- TRIB2
- Gene Name
- tribbles pseudokinase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021643.3 |
1937 |
Silent Mutation |
CAC,CAT |
H167H |
NP_067675.1 |
View Full Product Details