Product Details

SNP ID
rs112220361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:179104306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACCTTTACAAAATACAATACA[C/T]ATATTTTGATGATATATTCTGAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SESTD1 PubMed Links

Gene Details

Gene
SESTD1
Gene Name
SEC14 and spectrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178123.4 7834 UTR 3 NP_835224.3
XM_011512139.2 7834 UTR 3 XP_011510441.1
XM_011512141.2 7834 UTR 3 XP_011510443.1
XM_017005250.1 7834 UTR 3 XP_016860739.1
XM_017005251.1 7834 UTR 3 XP_016860740.1

View Full Product Details