Product Details

SNP ID
rs114023343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189772736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGTTAGTTTCTATTTAGGTCT[C/T]TGCAGTACGCAAGGAAAGTATTCTA
Phenotype
MIM: 609803 MIM: 610073
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKAR PubMed Links

Gene Details

Gene
ANKAR
Gene Name
ankyrin and armadillo repeat containing
There are no transcripts associated with this gene.

Gene
ORMDL1
Gene Name
ORMDL sphingolipid biosynthesis regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128150.1 Intron NP_001121622.1
NM_016467.4 Intron NP_057551.1
XM_005246970.2 Intron XP_005247027.1
XM_011512199.2 Intron XP_011510501.1
XM_011512200.2 Intron XP_011510502.1
XM_017005347.1 Intron XP_016860836.1
XM_017005348.1 Intron XP_016860837.1
Gene
OSGEPL1
Gene Name
O-sialoglycoprotein endopeptidase-like 1
There are no transcripts associated with this gene.

Gene
OSGEPL1-AS1
Gene Name
OSGEPL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details