Product Details

SNP ID
rs112626183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:217284550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTGAGTTCCTATAAACCAGTT[G/T]GAAAGGAATTACTGTCTGGAGGCAT
Phenotype
MIM: 608262
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DIRC3 PubMed Links

Gene Details

Gene
DIRC3
Gene Name
disrupted in renal carcinoma 3
There are no transcripts associated with this gene.

Gene
DIRC3-AS1
Gene Name
DIRC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details